siRNA Details
Gene Symbol: Ei24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025660.1 | NP_001020831.1 | 10 | 849 | ||
XM_006242800.2 | XP_006242862.1 | 9 | 940 | |||
XM_006242801.2 | XP_006242863.1 | 9 | 897 | |||
GenBank mRNA | BC098721.1 | - | 10 | 849 | 7370909 |
