siRNA Details
| Gene Symbol: Olr1198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000436.1 | NP_001000436.1 | 1 | 577 | ||
| Gene Symbol: Olr1197 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001120.1 | NP_001001120.1 | 1 | 577 | ||

Olr1198
Genomic Map