siRNA Details
| Gene Symbol: Olr1222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000440.1 | NP_001000440.1 | 1 | 157 | ||
| Gene Symbol: LOC103693050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766090.1 | XP_008764312.1 | 1 | 157 | ||

Olr1222
Genomic Map