siRNA Details
| Gene Symbol: Olr1241 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000449.1 | NP_001000449.1 | 1 | 520 | ||
| Gene Symbol: LOC100912488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008762662.1 | XP_008760884.1 | 1 | 520 | ||

Olr1241
Genomic Map