siRNA Details
| Gene Symbol: Olr1259 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000455.1 | NP_001000455.1 | 1 | 894 | ||
| Gene Symbol: LOC100912340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749617.3 | XP_003749665.2 | 1 | 992 | ||

Olr1259
Genomic Map