siRNA Details
| Gene Symbol: Olr1297 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000461.1 | NP_001000461.1 | 1 | 643 | ||
| Gene Symbol: Olr1301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000462.1 | NP_001000462.1 | 643 | |||
| XM_003750500.1 | XP_003750548.1 | 1 | 205 | |||

Olr1297
Genomic Map