siRNA Details
Gene Symbol: Olr1311 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000467.1 | NP_001000467.1 | 1 | 841 |
Gene Symbol: Olr1309 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000466.1 | NP_001000466.1 | 1 | 841 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
