siRNA Details
| Gene Symbol: Olr1320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000471.1 | NP_001000471.1 | 1 | 106 | ||
| Gene Symbol: Olr1318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001087.2 | NP_001001087.2 | 1 | 106 | ||
More Information
Set Membership: |
|

Olr1320
Genomic Map