siRNA Details
| Gene Symbol: Olr1325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000473.1 | NP_001000473.1 | 1 | 436 | ||
| Gene Symbol: Olr1328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000599.1 | NP_001000599.1 | 1 | 436 | ||

Olr1325
Genomic Map