siRNA Details
| Gene Symbol: Olr1326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000474.1 | NP_001000474.1 | 1 | 590 | ||
| Gene Symbol: Olr1332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000477.1 | NP_001000477.1 | 1 | 590 | ||

Olr1326
Genomic Map