siRNA Details
| Gene Symbol: Olr1332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000477.1 | NP_001000477.1 | 1 | 47 | ||
| Gene Symbol: Olr1331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000790.1 | NP_001000790.1 | 47 | |||

Olr1332
Genomic Map