siRNA Details
| Gene Symbol: Olr1335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000479.1 | NP_001000479.1 | 1 | 739 | ||
| Gene Symbol: Olr1334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000478.1 | NP_001000478.1 | 1 | 739 | ||

Olr1335
Genomic Map