siRNA Details
| Gene Symbol: Olr1340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750514.2 | XP_003750562.1 | 1 | 271 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr1340| Gene Symbol: Olr1340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750514.2 | XP_003750562.1 | 1 | 271 | ||