siRNA Details
| Gene Symbol: Crtam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106813.1 | NP_001100283.1 | 3 | 310 | ||
| XM_008766122.1 | XP_008764344.1 | 3 | 415 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crtam| Gene Symbol: Crtam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106813.1 | NP_001100283.1 | 3 | 310 | ||
| XM_008766122.1 | XP_008764344.1 | 3 | 415 | |||