siRNA Details
| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013953.1 | NP_001013975.1 | 5, 6 | 789 | ||
| XM_006242913.2 | XP_006242975.1 | 6, 7 | 1271 | |||
| GenBank mRNA | BC089910.1 | - | 5, 6 | 789 | 7304742 | |

Ccdc153
Genomic Map