siRNA Details
| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013953.1 | NP_001013975.1 | 3 | 498 | ||
| XM_006242913.2 | XP_006242975.1 | 4 | 980 | |||
| GenBank mRNA | BC089910.1 | - | 3 | 498 | 7304742 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc153| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013953.1 | NP_001013975.1 | 3 | 498 | ||
| XM_006242913.2 | XP_006242975.1 | 4 | 980 | |||
| GenBank mRNA | BC089910.1 | - | 3 | 498 | 7304742 | |