siRNA Details
| Gene Symbol: Abcg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106816.1 | NP_001100286.1 | 3 | 657 | ||
| XM_006242915.2 | XP_006242977.1 | 3 | 6389 | |||
| XM_006242916.2 | XP_006242978.1 | 3 | 747 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcg4| Gene Symbol: Abcg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106816.1 | NP_001100286.1 | 3 | 657 | ||
| XM_006242915.2 | XP_006242977.1 | 3 | 6389 | |||
| XM_006242916.2 | XP_006242978.1 | 3 | 747 | |||