siRNA Details
| Gene Symbol: Bcl9l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106817.1 | NP_001100287.1 | 11 | 4602 | ||
| XM_006242923.2 | XP_006242985.1 | 10 | 6317 | |||
| XM_006242924.2 | XP_006242986.1 | 9 | 6260 | |||
| XM_006242925.2 | XP_006242987.1 | 9 | 6215 | |||
| XM_006242928.2 | XP_006242990.1 | 7 | 3940 | |||

Bcl9l
Genomic Map