siRNA Details
| Gene Symbol: Arcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007662.1 | NP_001007663.1 | 3, 4 | 573 | ||
| GenBank mRNA | BC081979.1 | - | 3, 4 | 573 | 7128203 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_145985, AK031385, AK028556, AK154066, BC024127, and others.

Arcn1
Genomic Map