siRNA Details
| Gene Symbol: Ift46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024760.1 | NP_001019931.1 | 8 | 848 | ||
| XM_006242930.2 | XP_006242992.1 | 9 | 987 | |||
| XM_006242931.2 | XP_006242993.1 | 8 | 1128 | |||
| GenBank mRNA | BC079388.1 | - | 8 | 754 | 7111667 | |
| BC097954.1 | - | 8 | 848 | 7456463 | ||

Ift46
Genomic Map