siRNA Details
| Gene Symbol: Cd3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077646.2 | NP_001071114.1 | 3 | 188 | ||
| GenBank mRNA | S79711.1 | - | 3 | 188 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd3g| Gene Symbol: Cd3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077646.2 | NP_001071114.1 | 3 | 188 | ||
| GenBank mRNA | S79711.1 | - | 3 | 188 | ||