siRNA Details
Gene Symbol: Bud13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025277.1 | NP_001020448.1 | 6, 7 | 1423 | ||
GenBank mRNA | BC097995.1 | - | 6, 7 | 1423 | 7446660 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bud13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025277.1 | NP_001020448.1 | 6, 7 | 1423 | ||
GenBank mRNA | BC097995.1 | - | 6, 7 | 1423 | 7446660 |