siRNA Details
| Gene Symbol: Rexo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008326.1 | NP_001008327.1 | 5 | 640 | ||
| XM_006243001.2 | XP_006243063.1 | 5 | 554 | |||
| GenBank mRNA | BC086420.1 | - | 5 | 640 | 7300740 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rexo2| Gene Symbol: Rexo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008326.1 | NP_001008327.1 | 5 | 640 | ||
| XM_006243001.2 | XP_006243063.1 | 5 | 554 | |||
| GenBank mRNA | BC086420.1 | - | 5 | 640 | 7300740 | |