siRNA Details
| Gene Symbol: Cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015010.1 | NP_001015010.1 | 5 | 597 | ||
| XM_006243071.2 | XP_006243133.1 | 4 | 540 | |||
| GenBank mRNA | BC092630.1 | - | 5 | 597 | 7314828 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cib2| Gene Symbol: Cib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015010.1 | NP_001015010.1 | 5 | 597 | ||
| XM_006243071.2 | XP_006243133.1 | 4 | 540 | |||
| GenBank mRNA | BC092630.1 | - | 5 | 597 | 7314828 | |