siRNA Details
| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106823.1 | NP_001100293.1 | 5 | 1364 | ||
| XM_006243072.2 | XP_006243134.1 | 5 | 1159 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hykk| Gene Symbol: Hykk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106823.1 | NP_001100293.1 | 5 | 1364 | ||
| XM_006243072.2 | XP_006243134.1 | 5 | 1159 | |||