siRNA Details
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005547.1 | NP_001005547.1 | 5 | 546 | ||
GenBank mRNA | BC082102.1 | - | 5 | 546 | 7134588 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005547.1 | NP_001005547.1 | 5 | 546 | ||
GenBank mRNA | BC082102.1 | - | 5 | 546 | 7134588 |