siRNA Details
| Gene Symbol: Mpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004081.1 | NP_001004081.1 | 5 | 652 | ||
| GenBank mRNA | BC079111.1 | - | 5 | 652 | 7125738 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpi| Gene Symbol: Mpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004081.1 | NP_001004081.1 | 5 | 652 | ||
| GenBank mRNA | BC079111.1 | - | 5 | 652 | 7125738 | |