siRNA Details
| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013108.3 | NP_001013126.2 | 3, 4 | 846 | ||
| GenBank mRNA | BC083881.1 | - | 3, 4 | 846 | 7131418 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arih1| Gene Symbol: Arih1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013108.3 | NP_001013126.2 | 3, 4 | 846 | ||
| GenBank mRNA | BC083881.1 | - | 3, 4 | 846 | 7131418 | |