siRNA Details
| Gene Symbol: Parp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106828.1 | NP_001100298.1 | 10 | 1042 | ||
| XM_006243191.1 | XP_006243253.1 | 10 | 1323 | |||
| XM_006243192.2 | XP_006243254.1 | 9 | 1981 | |||
| XM_006243193.2 | XP_006243255.1 | 9 | 850 | |||
| XM_008766231.1 | XP_008764453.1 | 9 | 1981 | |||
| XM_008766232.1 | XP_008764454.1 | 9 | 1981 | |||
| XM_008766233.1 | XP_008764455.1 | 9 | 1981 | |||
| GenBank mRNA | BC167025.1 | - | 8 | 577 | 7365328 | |
| FQ223826.1 | - | 9 | 1705 | |||

Parp6
Genomic Map