siRNA Details
| Gene Symbol: Gramd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243197.2 | XP_006243259.1 | 5 | 615 | ||
| XM_006243198.2 | XP_006243260.1 | 6 | 976 | |||
| XM_006243199.2 | XP_006243261.1 | 5 | 1024 | |||
| XM_006243200.2 | XP_006243262.1 | 5 | 1364 | |||
| XM_006243201.2 | XP_006243263.1 | 5 | 2825 | |||
| XM_008766374.1 | XP_008764596.1 | 5 | 495 | |||
| XM_217153.7 | XP_217153.5 | 5 | 414 | |||

Gramd2
Genomic Map