siRNA Details
| Gene Symbol: SLC30A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004434.2 | NP_001004434.1 | 1, 2 | 300 | ||
| NM_032513.4 | NP_115902.1 | 1, 2 | 300 | |||
| GenBank mRNA | AF370409.1 | - | 1, 2 | 147 | ||
| AK094027.1 | - | 1, 2 | 116 | |||
| BC006251.2 | - | 1, 2 | 275 | 3948165 | ||
| DR004258.1 | - | 1, 2 | 307 | |||

SLC30A2
Genomic Map