siRNA Details
| Gene Symbol: Iqch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106830.1 | NP_001100300.1 | 7 | 793 | ||
| XM_008766234.1 | XP_008764456.1 | 2 | 348 | |||
| XM_008766235.1 | XP_008764457.1 | 2 | 348 | |||
| XM_008766236.1 | XP_008764458.1 | 2 | 348 | |||
| XM_008766237.1 | XP_008764459.1 | 7 | 849 | |||

Iqch
Genomic Map