siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007663.1 | NP_001007664.1 | 8 | 1234 | ||
| XM_008766238.1 | XP_008764460.1 | 9 | 1536 | |||
| XM_008766239.1 | XP_008764461.1 | 10 | 1488 | |||
| GenBank mRNA | BC082006.1 | - | 8 | 1234 | 7131624 | |
| FQ231531.1 | - | 4 | 314 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_175153, NM_001077631, BC024913, AK169772, AK032095, and others.

Vwa9
Genomic Map