siRNA Details
| Gene Symbol: Vwa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007663.1 | NP_001007664.1 | 5 | 919 | ||
| XM_008766238.1 | XP_008764460.1 | 6 | 1221 | |||
| XM_008766239.1 | XP_008764461.1 | 7 | 1173 | |||
| GenBank mRNA | BC082006.1 | - | 5 | 919 | 7131624 | |

Vwa9
Genomic Map