siRNA Details
| Gene Symbol: Clpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007803.1 | NP_001007804.1 | 6 | 704 | ||
| GenBank mRNA | BC085867.1 | - | 6 | 830 | 7190926 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011802, AF134983, AK087848.

Clpx
Genomic Map