siRNA Details
| Gene Symbol: SLC30A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013309.5 | NP_037441.2 | 6 | 1269 | ||
| XM_011521997.2 | XP_011520299.1 | 6 | 1773 | |||
| XM_017022560.1 | XP_016878049.1 | 5 | 1075 | |||
| GenBank mRNA | AF025409.1 | - | 5 | 957 | ||
| AK290874.1 | - | 5 | 1075 | |||
| AY029482.1 | - | 2 | 234 | |||
| BC026089.2 | - | 6 | 1269 | 4829839 | ||

SLC30A4
Genomic Map