siRNA Details
| Gene Symbol: Spg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006987.1 | NP_001006988.1 | 7 | 821 | ||
| XM_006243251.2 | XP_006243313.1 | 7 | 852 | |||
| GenBank mRNA | BC083760.1 | - | 7 | 821 | 7192933 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spg21| Gene Symbol: Spg21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006987.1 | NP_001006988.1 | 7 | 821 | ||
| XM_006243251.2 | XP_006243313.1 | 7 | 852 | |||
| GenBank mRNA | BC083760.1 | - | 7 | 821 | 7192933 | |