siRNA Details
| Gene Symbol: Ns5atp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201418.1 | NP_958821.1 | 2, 3 | 124 | ||
| GenBank mRNA | AY496944.1 | - | 2, 3 | 124 | ||
| FQ220183.1 | - | 2, 3 | 201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ns5atp9| Gene Symbol: Ns5atp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201418.1 | NP_958821.1 | 2, 3 | 124 | ||
| GenBank mRNA | AY496944.1 | - | 2, 3 | 124 | ||
| FQ220183.1 | - | 2, 3 | 201 | |||