siRNA Details
| Gene Symbol: Fam96a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008327.1 | NP_001008328.1 | 5 | 531 | ||
| GenBank mRNA | BC086524.1 | - | 5 | 531 | 7303322 | |
| FQ215407.1 | - | 5 | 568 | |||
| FQ216458.1 | - | 5 | 569 | |||
| FQ218392.1 | - | 5 | 512 | |||
| FQ218618.1 | - | 5 | 575 | |||
| FQ220244.1 | - | 5 | 491 | |||
| FQ225271.1 | - | 5 | 428 | |||
| FQ232448.1 | - | 5 | 529 | |||
| FQ233915.1 | - | 5 | 436 | |||

Fam96a
Genomic Map