siRNA Details
| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106833.1 | NP_001100303.1 | 6 | 1368 | ||
| XM_008766242.1 | XP_008764464.1 | 5 | 1185 | |||
| GenBank mRNA | FQ230750.1 | - | 3 | 357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lactb| Gene Symbol: Lactb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106833.1 | NP_001100303.1 | 6 | 1368 | ||
| XM_008766242.1 | XP_008764464.1 | 5 | 1185 | |||
| GenBank mRNA | FQ230750.1 | - | 3 | 357 | ||