siRNA Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106836.1 | NP_001100306.1 | 4 | 1330 | ||
| XM_006243372.2 | XP_006243434.1 | 4 | 1483 | |||
| XM_006243373.2 | XP_006243435.1 | 4 | 1482 | |||
| XM_006243374.2 | XP_006243436.1 | 4 | 1482 | |||
| XM_006243376.2 | XP_006243438.1 | 5 | 1575 | |||
| XM_006243377.2 | XP_006243439.1 | 4 | 1512 | |||
| XM_008766243.1 | XP_008764465.1 | 4 | 1482 | |||
| XM_008766244.1 | XP_008764466.1 | 4 | 1261 | |||
| XM_008766245.1 | XP_008764467.1 | 4 | 1483 | |||

Rnf111
Genomic Map