siRNA Details
| Gene Symbol: Tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004248.1 | NP_001004248.1 | 5 | 997 | ||
| XM_006243435.2 | XP_006243497.1 | 6 | 1107 | |||
| GenBank mRNA | BC079203.1 | - | 5 | 997 | 7131766 | |
| FQ209432.1 | - | 2 | 187 | |||

Tmem30a
Genomic Map