siRNA Details
| Gene Symbol: Tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004248.1 | NP_001004248.1 | 3 | 655 | ||
| XM_006243435.2 | XP_006243497.1 | 4 | 765 | |||
| GenBank mRNA | BC079203.1 | - | 3 | 655 | 7131766 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem30a| Gene Symbol: Tmem30a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004248.1 | NP_001004248.1 | 3 | 655 | ||
| XM_006243435.2 | XP_006243497.1 | 4 | 765 | |||
| GenBank mRNA | BC079203.1 | - | 3 | 655 | 7131766 | |