siRNA Details
| Gene Symbol: Senp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106842.2 | NP_001100312.2 | 9 | 1141 | ||
| XM_006243436.1 | XP_006243498.1 | 8 | 1496 | |||
| XM_006243437.1 | XP_006243499.1 | 7 | 800 | |||
| GenBank mRNA | FQ213152.1 | - | 6 | 714 | ||

Senp6
Genomic Map