siRNA Details
| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277283.1 | NP_001264212.1 | 4 | 644 | ||
| GenBank mRNA | BC159433.1 | - | 4 | 636 | 9093094 | |
| FQ215646.1 | - | 4 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irak1bp1| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277283.1 | NP_001264212.1 | 4 | 644 | ||
| GenBank mRNA | BC159433.1 | - | 4 | 636 | 9093094 | |
| FQ215646.1 | - | 4 | 578 | |||