siRNA Details
| Gene Symbol: Plod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ430860.1 | - | 8, 9 | 1139 | ||
| AJ430861.1 | - | 8, 9 | 1139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plod2| Gene Symbol: Plod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ430860.1 | - | 8, 9 | 1139 | ||
| AJ430861.1 | - | 8, 9 | 1139 | |||