siRNA Details
| Gene Symbol: Il1rapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166342.1 | NP_001159814.1 | 11 | 2780 | ||
| XM_008773406.1 | XP_008771628.1 | 10 | 2045 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il1rapl2| Gene Symbol: Il1rapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166342.1 | NP_001159814.1 | 11 | 2780 | ||
| XM_008773406.1 | XP_008771628.1 | 10 | 2045 | |||