siRNA Details
| Gene Symbol: Xrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006243594.2 | XP_006243656.1 | 35 | 5076 | ||
| XM_006243596.2 | XP_006243658.1 | 35 | 5076 | |||
| XM_006243597.2 | XP_006243659.1 | 35 | 5061 | |||
| XM_006243598.2 | XP_006243660.1 | 35 | 4986 | |||
| XM_006243599.2 | XP_006243661.1 | 34 | 4962 | |||
| XM_217233.8 | XP_217233.5 | 35 | 5061 | |||

Xrn1
Genomic Map