siRNA Details
| Gene Symbol: Spsb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106849.2 | NP_001100319.1 | 2 | 1243 | ||
| GenBank mRNA | BC168912.1 | - | 2 | 1243 | 7191741 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spsb4| Gene Symbol: Spsb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106849.2 | NP_001100319.1 | 2 | 1243 | ||
| GenBank mRNA | BC168912.1 | - | 2 | 1243 | 7191741 | |