siRNA Details
| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100547.1 | NP_001094017.1 | 3 | 1014 | ||
| GenBank mRNA | X94186.1 | - | 3 | 597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp7| Gene Symbol: Dusp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100547.1 | NP_001094017.1 | 3 | 1014 | ||
| GenBank mRNA | X94186.1 | - | 3 | 597 | ||